Abnova
Manufacturer Code:PAB5542
Catalog # 89-131-895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human RAB8A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RAB8A.
For Research Use Only
Protein Aliases: MEL; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); Oncogene c-mel; RAB8; ras-associated protein RAB8; Ras-related protein Rab-8A
Gene Aliases: MEL; RAB8; RAB8A
UniProt ID: (Human) P61006
Entrez Gene ID: (Human) 4218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.