Abnova
Manufacturer Code:PAB24746
Catalog # 89-126-412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to RAB7A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RAB7A.
For Research Use Only
Protein Aliases: FLJ20819; PRO2706; RAB7; RAB7, member RAS oncogene family; Ras-associated protein RAB7; Ras-related protein Rab-7a
Gene Aliases: PRO2706; RAB7; RAB7A
UniProt ID: (Human) P51149
Entrez Gene ID: (Human) 7879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.