Invitrogen
Manufacturer Code:PA523138
Catalog # PIPA523138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (97 NQYVNKKFSNQYKA 110) of Rab-7 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein (rab7); PRO2706 RAB7 RAB7 member RAS oncogene family Ras-associated protein RAB7 ras-related protein Rab-7a; RAB7, member RAS oncogene family; Ras-associated protein RAB7; Ras-related protein BRL-RAS; Ras-related protein p23; Ras-related protein Rab-7a
Gene Aliases: PRO2706; RAB7; RAB7A
UniProt ID: (Dog) P18067, (Human) P51149, (Bovine) Q3T0F5, (Mouse) P51150, (Rat) P09527
Entrez Gene ID: (Chicken) 416016, (Dog) 404007, (Human) 7879, (Bovine) 509970, (Mouse) 19349, (Rat) 29448
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.