Novus Biologicals
Manufacturer Code:NBP288116
Catalog # NB035738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human RAB6A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAB6A&prime GTP-binding protein RAB6B Oncogene RAB6 Rab GTPase Rab-6 RAB6 member RAS oncogene family RAB6A&prime Rab6A variant 3 RAB6A member RAS oncogene family RAB6B RAB6rab-6 ras-related protein Rab-6A; Rab GTPase; Rab-6; RAB6, member RAS oncogene family; Ras-related protein Rab-6A
Gene Aliases: RAB6; RAB6A
UniProt ID: (Human) P20340
Entrez Gene ID: (Human) 5870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.