Proteintech
Manufacturer Code:101872AP150UL
Catalog # 50-172-6092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB6A Fusion Protein Ag0240 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Rab 6 RAB6 RAB6A RAB6A&prime RAB6B Ras related protein Rab 6A; Rab GTPase; Rab-6; RAB6, member RAS oncogene family; Ras-related protein Rab-6A
Gene Aliases: 2610028L11Rik; AA419671; MNCb-1660; RAB6; RAB6A
UniProt ID: (Human) P20340, (Mouse) P35279
Entrez Gene ID: (Human) 5870, (Mouse) 19346
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.