Invitrogen
Manufacturer Code:MA524745
Catalog # PIMA524745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1616CT314.65.31 |
Immunogen | human RAB5C recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: L1880; RAB5C, member of RAS oncogene family; RAB5L; Ras-related protein Rab-5C; Ras-related protein Rab-5C L1880 RAB5L RAB5C RABL
Gene Aliases: L1880; RAB5C; RAB5CL; RAB5L; RABL
UniProt ID: (Human) P51148
Entrez Gene ID: (Human) 5878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.