Invitrogen
Manufacturer Code:PA570358
Catalog # PIPA570358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human RAB5C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L1880; L1880 RAB member of RAS oncogene family like RAB5C RAB5C member of RAS oncogene family RAB5C member RAS oncogene family RAB5C RAB5CL RAB5L RABL Ras-related protein Rab-5C; RAB5B, member RAS oncogene family; RAB5C; RAB5C, member of RAS oncogene family; RAB5L; Ras-related protein Rab-5C; small GTPase
Gene Aliases: AI326010; L1880; RAB5B; RAB5C; RAB5CL; RAB5L; RABL
UniProt ID: (Dog) P51147, (Human) P51148, (Bovine) Q58DS9, (Mouse) P35278
Entrez Gene ID: (Dog) 403941, (Pig) 100523862, (Human) 5878, (Bovine) 613749, (Rabbit) 100348021, (Horse) 100053004, (Mouse) 19345, (Rat) 287709, (Guinea pig) 100716247
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.