Novus Biologicals
Manufacturer Code:NBP29305120UL
Catalog # NB027996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human RAB5C (NP_958842.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L1880; L1880 MGC117217 RAB5C member of RAS oncogene family RAB5C member RAS oncogene family RAB5CL RAB5L RABLMGC138857 ras-related protein Rab-5C; RAB5C, member of RAS oncogene family; RAB5L; Ras-related protein Rab-5C
Gene Aliases: L1880; RAB5C; RAB5CL; RAB5L; RABL
UniProt ID: (Human) P51148
Entrez Gene ID: (Human) 5878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.