Proteintech
Manufacturer Code:272191AP150UL
Catalog # 50-173-5608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB5C Fusion Protein Ag25849 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L1880; L1880 RAB5C RAB5CL RAB5L RABL Ras related protein Rab 5C; RAB5C, member of RAS oncogene family; RAB5L; Ras-related protein Rab-5C
Gene Aliases: L1880; RAB5C; RAB5CL; RAB5L; RABL
UniProt ID: (Human) P51148
Entrez Gene ID: (Human) 5878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.