Proteintech
Manufacturer Code:666541IG150UL
Catalog # 50-173-7072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B2A8 |
Immunogen | RAB43 Fusion Protein Ag26911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ISY1 RAB11B RAB41 RAB43 Ras related protein Rab 41 Ras related protein Rab 43; Ras-related protein Rab-41; Ras-related protein Rab-43
Gene Aliases: RAB11B; RAB41; RAB43
UniProt ID: (Human) Q86YS6
Entrez Gene ID: (Human) 339122
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.