OriGene
Manufacturer Code:TA808961
Catalog # 50-168-2491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D1 |
Immunogen | Full length human recombit protein of human RAB3IP produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Rab-3A-interacting protein; Rab3A-interacting protein; RAB3IP; Rabin-3; RABIN3 RABIN8; SSX2 interacting protein; SSX2-interacting protein
Gene Aliases: RAB3IP; RABIN3; RABIN8
UniProt ID: (Human) Q96QF0
Entrez Gene ID: (Human) 117177
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.