Invitrogen
Manufacturer Code:PA544021
Catalog # PIPA544021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RAB3IP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gene trap PAT 12; Rab-3A-interacting protein; RAB3A interacting protein (rabin3); RAB3A-interacting protein; RAB3IP; rab3ip {ECO:0000250|UniProtKB:Q96QF0}; rabin 3; Rabin-3; SSX2 interacting protein; SSX2-interacting protein
Gene Aliases: B230311A06; Gtpat12; RAB3IP; RABIN3; RABIN8
UniProt ID: (Human) Q96QF0, (Rat) Q62739, (Mouse) Q68EF0
Entrez Gene ID: (Human) 117177, (Horse) 100052332, (Bovine) 533284, (Dog) 481159, (Rabbit) 100342115, (Rat) 29885, (Mouse) 216363
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.