Novus Biologicals
Manufacturer Code:NBP15674520UL
Catalog # NBP15674520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RAB3IP(RAB3A interacting protein (rabin3)) The peptide sequence was selected from the N terminal of RAB3IP. Peptide sequence APCSTSGVTAGLTKLTTRKDNYNAEREFLQGATITEACDGSDDIFGLSTD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14660 FLJ22548 MGC71495 RAB3A interacting protein (rabin3) rab-3A-interacting protein Rab3A-interacting protein rabin-3 RABIN3 SSX2 interacting protein SSX2-interacting protein; Rab-3A-interacting protein; Rab3A-interacting protein; RAB3IP; Rabin-3; SSX2 interacting protein; SSX2-interacting protein
Gene Aliases: RAB3IP; RABIN3; RABIN8
UniProt ID: (Human) Q96QF0
Entrez Gene ID: (Human) 117177
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.