Invitrogen
Manufacturer Code:PA539386
Catalog # PIPA539386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human RAB3GAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAB3 GTPase activating protein subunit 2 (non-catalytic); Rab3 GTPase-activating protein 150 kDa subunit; Rab3 GTPase-activating protein non-catalytic subunit; Rab3-GAP p150; Rab3-GAP regulatory subunit; RAB3-GAP150; RAB3-GAP150 RAB3GAP150 WARBM2 p150 RGAP-iso rab3 GTPase-activating protein 150 kDa subunit rab3 GTPase-activating protein non-catalytic subunit rab3-GAP p150 rab3-GAP regulatory subunit; RGAP-iso
Gene Aliases: 1110059F07Rik; 2010002H18Rik; 5830469C09; AI851069; AW743433; KIAA0839; mKIAA0839; p150; RAB3-GAP150; RAB3GAP150; RAB3GAP2; RGD1311518; SPG69; WARBM2
UniProt ID: (Human) Q9H2M9, (Mouse) Q8BMG7, (Rat) Q5U1Z0
Entrez Gene ID: (Human) 25782, (Mouse) 98732, (Rat) 289350
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.