Abnova
Manufacturer Code:PAB21794
Catalog # 89-123-496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human RAB3GAP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant RAB3GAP2.
For Research Use Only
Protein Aliases: DKFZp434D245; FLJ14579; KIAA0839; p150; RAB3 GTPase activating protein subunit 2 (non-catalytic); Rab3 GTPase-activating protein 150 kDa subunit; Rab3 GTPase-activating protein non-catalytic subunit; Rab3-GAP p150; Rab3-GAP regulatory subunit; RAB3-GAP150; RAB3GAP150; RGAP-iso
Gene Aliases: KIAA0839; p150; RAB3-GAP150; RAB3GAP150; RAB3GAP2; SPG69; WARBM2
UniProt ID: (Human) Q9H2M9
Entrez Gene ID: (Human) 25782
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.