Novus Biologicals
Manufacturer Code:NBP2854470.1ML
Catalog # NB033376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human RAB3GAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0066P130 RAB3 GTPase activating protein subunit 1 (catalytic) RAB3 GTPase-activating protein 130 kDa subunit rab3 GTPase-activating protein catalytic subunit Rab3-GAP Rab3-GAP p130 RAB3GAPDKFZp434A012 WARBM1RAB3GAP130; RAB3 GTPase activating protein subunit 1 (catalytic); RAB3 GTPase-activating protein 130 kDa subunit; Rab3 GTPase-activating protein catalytic subunit; Rab3-GAP; Rab3-GAP p130
Gene Aliases: KIAA0066; P130; RAB3GAP; RAB3GAP1; RAB3GAP130; WARBM1
UniProt ID: (Human) Q15042
Entrez Gene ID: (Human) 22930
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.