Proteintech
Manufacturer Code:123201AP150UL
Catalog # 50-172-7889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB3D Fusion Protein Ag2973 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D2 2 GOV RAB16 RAB3D RAD3D Ras related protein Rab 3D; glioblastoma overexpressed; GTP-binding protein Rab-3D; Rab3D upregulated with myeloid differentiation; Ras-related protein Rab-3D
Gene Aliases: C130057E11Rik; D2-2; GOV; RAB16; RAB3D; RAD3D
UniProt ID: (Human) O95716, (Mouse) P35276, (Rat) Q63942
Entrez Gene ID: (Human) 9545, (Mouse) 19340, (Rat) 140665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.