Novus Biologicals
Manufacturer Code:NBP220042
Catalog # NBP220042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human RAB35. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAY; GTP-binding protein RAY H-ray RAB1C RAB35 member RAS oncogene family Ras-related protein Rab-1C ras-related protein rab-1c (GTP-binding protein ray) ras-related protein Rab-35 RAY; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35
Gene Aliases: H-ray; RAB1C; RAB35; RAY
UniProt ID: (Human) Q15286
Entrez Gene ID: (Human) 11021
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.