Proteintech
Manufacturer Code:113292AP150UL
Catalog # 50-557-073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB35 Fusion Protein Ag1872 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein RAY H ray RAB1C RAB35 Ras related protein Rab 1C Ras related protein Rab 35 RAY; GTP-binding protein RAY; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35
Gene Aliases: 9530019H02Rik; AU040256; H-ray; RAB1C; RAB35; RAY
UniProt ID: (Human) Q15286, (Mouse) Q6PHN9, (Rat) Q5U316
Entrez Gene ID: (Human) 11021, (Mouse) 77407, (Rat) 288700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.