Invitrogen
Manufacturer Code:PA539401
Catalog # PIPA539401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human RAB31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein Rab0; monomeric GTP-binding protein; RAB22B; Ras-related protein Rab-22B; Ras-related protein Rab-31
Gene Aliases: 1700093E07Rik; AI415285; Rab0; RAB22B; RAB31
UniProt ID: (Human) Q13636, (Rat) Q6GQP4, (Mouse) Q921E2
Entrez Gene ID: (Human) 11031, (Rat) 246324, (Mouse) 106572
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.