Novus Biologicals
Manufacturer Code:NBP297073UV
Catalog # NB080946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant RAB31 (NP_006859.2 Met1-Cys195). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Rab22B RAB31 member RAS oncogene family Ras-related protein Rab-22B ras-related protein Rab-31; Ras-related protein Rab-22B; Ras-related protein Rab-31
Gene Aliases: RAB22B; RAB31
UniProt ID: (Human) Q13636
Entrez Gene ID: (Human) 11031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.