OriGene
Manufacturer Code:TA505341
Catalog # 50-167-5835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F4 |
Immunogen | Full length human recombit protein of human RAB30 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: RAB30 member RAS oncogene family; Ras-related protein Rab-30
Gene Aliases: 5033421K01Rik; AI323892; RAB30; Rsb30
UniProt ID: (Human) Q15771, (Mouse) Q923S9
Entrez Gene ID: (Dog) 476787, (Human) 27314, (Rat) 308821, (Mouse) 75985
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.