Proteintech
Manufacturer Code:134121AP150UL
Catalog # 50-555-531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB27B Fusion Protein Ag4064 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C25KG; C25KG RAB27B Ras related protein Rab 27B; Ras-related protein Rab-27B
Gene Aliases: 2310021G14Rik; B130064M09Rik; C25KG; RAB27B
UniProt ID: (Human) O00194, (Mouse) Q99P58, (Rat) Q99P74
Entrez Gene ID: (Human) 5874, (Mouse) 80718, (Rat) 84590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.