Abnova
Manufacturer Code:H00005873M02J
Catalog # 89-335-109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G7 |
Immunogen | RAB27A (NP_004571 122 a.a. &sim 221 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: GS2 HsT18676 MGC117246 RAB27 RAM; GTP-binding protein Ram; mutant Ras-related protein Rab-27A; Rab-27; Ras-related protein Rab-27A
Gene Aliases: GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159
Entrez Gene ID: (Human) 5873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.