Invitrogen
Manufacturer Code:PA5102522
Catalog # PIPA5102522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RAB27A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210402C08Rik 2410003M20Rik 4933437C11Rik ash ashen GS2 GTP-binding protein Ram HsT18676 low Mr GTP-binding protein mutant Ras-related protein Rab-27A Rab27 rab-27 Rab27a RAB27A member RAS oncogene family RAM Ram p25 ras-related protein Rab-27A; ashen; GTP-binding protein Ram; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; Rab-27; Ram p25; Ras-related protein Rab-27A
Gene Aliases: 2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159, (Mouse) Q9ERI2, (Rat) P23640
Entrez Gene ID: (Human) 5873, (Mouse) 11891, (Rat) 50645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.