Abnova
Manufacturer Code:PAB28415
Catalog # 89-129-921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human RAB27A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4 °C. For long term storage,store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant RAB27A.
For Research Use Only
Protein Aliases: GS2; GTP-binding protein Ram; HsT18676; MGC117246; mutant Ras-related protein Rab-27A; Rab-27; RAB27; RAM; Ras-related protein Rab-27A
Gene Aliases: GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159
Entrez Gene ID: (Human) 5873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.