Invitrogen
Manufacturer Code:PA5110256
Catalog # PIPA5110256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-213 of human RAB25 (NP_065120.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV012978 CATX8 CATX-8 RAB11C RAB25 Ras-related protein Rab-25; CATX-8; Ras-related protein Rab-25
Gene Aliases: AV012978; CATX-8; CATX8; RAB11C; RAB25
UniProt ID: (Human) P57735, (Mouse) Q9WTL2
Entrez Gene ID: (Human) 57111, (Mouse) 53868, (Rat) 310632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.