Invitrogen
Manufacturer Code:PA544777
Catalog # PIPA544777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RAB22A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAB22A; Rab-14; Rab-22; RAB22, member RAS oncogene family; RAB22A member RAS oncogene family MGC16770; RAS-related protein RAB-22A
Gene Aliases: 3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; im:7136651; RAB22; RAB22A; zgc:101631
UniProt ID: (Human) Q9UL26, (Dog) P51154, (Mouse) P35285
Entrez Gene ID: (Human) 57403, (Horse) 100056212, (Dog) 403864, (Rabbit) 100350969, (Bovine) 617385, (Pig) 100144535, (Rat) 366265, (Mouse) 19334, (Guinea pig) 100731614, (Zebrafish) 100001531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.