Proteintech
Manufacturer Code:161811AP150UL
Catalog # 50-173-0775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein RAB22A; Rab 22 RAB22 RAB22A Ras related protein Rab 22A; Rab-14; Rab-22; RAB22, member RAS oncogene family; Ras-related protein Rab-22A
Gene Aliases: 3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; RAB22; RAB22A
UniProt ID: (Human) Q9UL26, (Mouse) P35285
Entrez Gene ID: (Human) 57403, (Mouse) 19334
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.