Invitrogen
Manufacturer Code:MA537710
Catalog # PIMA537710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1673CT571.17.71 |
Immunogen | recombinant protein between 1-201 amino acids from human RAB1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110011F09Rik RAB1B RAB1B member RAS oncogene family Rab1bl RAB1B-like member RAS oncogene family Ras-related protein Rab-1B small GTP-binding protein; RAB1B-like, member RAS oncogene family; Ras-related protein Rab-1B; small GTP-binding protein
Gene Aliases: 1110011F09Rik; RAB1B; Rab1bl
UniProt ID: (Human) Q9H0U4, (Mouse) Q9D1G1
Entrez Gene ID: (Human) 81876, (Rat) 100126191, (Mouse) 76308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.