Invitrogen
Manufacturer Code:PA577783
Catalog # PIPA577783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the C-terminal of Human RAB1B (aa 179-189) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAB1B-like, member RAS oncogene family; Ras-related protein Rab-1B; Ras-related protein Rab-1B RAB1B; small GTP-binding protein
Gene Aliases: RAB1B; Rab1bl
UniProt ID: (Human) Q9H0U4
Entrez Gene ID: (Human) 81876, (Rat) 100126191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.