Invitrogen
Manufacturer Code:PA544578
Catalog # PIPA544578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RAB1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein Rab1a; Rab GTPase YPT1 homolog; rab1; RAB1, member RAS oncogene family; RAB1A member RAS oncogene family DKFZP564B163 RAB1 YPT1; RAB1A-like protein; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1-related protein
Gene Aliases: Ac2-048; Gtbp; mKIAA3012; Rab-1; RAB1; RAB1A; Rab1r; YPT1
UniProt ID: (Human) P62820, (Dog) P62822, (Mouse) P62821, (Rat) Q6NYB7
Entrez Gene ID: (Bovine) 539339, (Rabbit) 100339709, (Goat) 100860796, (Human) 5861, (Dog) 403774, (Horse) 100051331, (Mouse) 19324, (Rat) 81754, (Guinea pig) 100719957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.