Invitrogen
Manufacturer Code:PA5110254
Catalog # PIPA5110254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RAB1A (NP_004152.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ac2-048 BOS_11764 DKFZP564B163 Gtbp mKIAA3012 RAB1 Rab-1 RAB1A Rab1r Ras-related protein Rab-1A Ypt1 YPT1-related protein; GTP binding protein Rab1a; Rab GTPase YPT1 homolog; RAB1, member RAS oncogene family; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1-related protein
Gene Aliases: Gtbp; mKIAA3012; Rab-1; RAB1; RAB1A; YPT1
UniProt ID: (Human) P62820, (Mouse) P62821
Entrez Gene ID: (Human) 5861, (Mouse) 19324
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.