Invitrogen
Manufacturer Code:PA595323
Catalog # PIPA595323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Rab18 identical to the related mouse sequence and different from the related rat sequence by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAB18 small GTPase; Ras-related protein Rab-18; Ras-related protein Rab-18 RAB18
Gene Aliases: RAB18; RAB18LI1; WARBM3
UniProt ID: (Human) Q9NP72, (Rat) Q5EB77
Entrez Gene ID: (Human) 22931, (Rat) 307039
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.