Invitrogen
Manufacturer Code:PA539400
Catalog # PIPA539400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human RAB18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein Rab18; RAB18 small GTPase; RAB18LI1 WARBM3 RAB18 small GTPase ras-related protein Rab-18; Ras-related protein Rab-18
Gene Aliases: AA959686; RAB18; RAB18LI1; WARBM3
UniProt ID: (Human) Q9NP72, (Mouse) P35293, (Rat) Q5EB77
Entrez Gene ID: (Human) 22931, (Mouse) 19330, (Rat) 307039
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.