Proteintech
Manufacturer Code:156621AP150UL
Catalog # 50-173-0385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB14 Fusion Protein Ag8092 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA165P4.3 (member RAS oncogene family); F protein-binding protein 1; FBP RAB 14 RAB14 Ras related protein Rab 14; GTPase Rab14; Ras-related protein Rab-14; small GTP binding protein RAB14
Gene Aliases: 0610030G24Rik; 2810475J17Rik; A830021G03Rik; AI314285; AI649155; D030017L14Rik; FBP; RAB-14; RAB14
UniProt ID: (Human) P61106, (Mouse) Q91V41, (Rat) P61107
Entrez Gene ID: (Human) 51552, (Mouse) 68365, (Rat) 94197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.