Invitrogen
Manufacturer Code:PA576509
Catalog # PIPA576509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human RAB13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; RAS-associated protein RAB13; Ras-related protein Rab-13
Gene Aliases: 0610007N03Rik; B230212B15Rik; GIG4; RAB13
UniProt ID: (Human) Q5U0A6, (Mouse) Q9DD03, (Rat) P35286
Entrez Gene ID: (Human) 5872, (Mouse) 68328, (Rat) 81756
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.