Invitrogen
Manufacturer Code:PA568292
Catalog # PIPA568292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human RAB11FIP5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gaf-1; GAF1 RIP11 pp75 gaf-1 gamma-SNAP-associated factor 1 phosphoprotein pp75 rab11 family-interacting protein 5 rab11-FIP5 rab11-interacting protein Rip11; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: 9130206P09Rik; C75969; D6Ertd32e; GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6, (Mouse) Q8R361
Entrez Gene ID: (Human) 26056, (Rhesus monkey) 713953, (Rat) 312502, (Mouse) 52055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.