Invitrogen
Manufacturer Code:PA557642
Catalog # PIPA557642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RAB11FIP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gaf-1; GAF1 RIP11 pp75 gaf-1 gamma-SNAP-associated factor 1 phosphoprotein pp75 rab11 family-interacting protein 5 rab11-FIP5 rab11-interacting protein Rip11; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6
Entrez Gene ID: (Human) 26056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.