Invitrogen
Manufacturer Code:PA5115815
Catalog # PIPA5115815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RAB11FIP5 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130206P09Rik C75969 D6Ertd32e GAF1 Gaf-1 Gamma-SNAP-associated factor 1 KIAA0857 Phosphoprotein pp75 pp75 RAB11 family interacting protein 5 RAB11 family interacting protein 5 (class I) rab11 family-interacting protein 5 RAB11FIP5 Rab11-FIP5 rab11-interacting protein Rip11 Rip11; Gaf-1; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: 9130206P09Rik; C75969; D6Ertd32e; GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6, (Mouse) Q8R361
Entrez Gene ID: (Human) 26056, (Rat) 312502, (Mouse) 52055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.