Abnova
Manufacturer Code:H00026056K
Catalog # 89-202-143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RAB11FIP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RAB11FIP5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp434H018 GAF1 KIAA0857 RIP11 pp75; Gaf-1; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6
Entrez Gene ID: (Human) 26056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.