Novus Biologicals
Manufacturer Code:NBP273764AF350
Catalog # NP273764A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI13B5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 (NP_116321) produced in E.coli. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ARFO2 Arfophilin-2 FIP4-Rab11 FLJ00131 KIAA1821arfophilin-2 MGC11316 MGC126566 RAB11 family interacting protein 4 (class II) rab11 family-interacting protein 4 Rab11-FIP4; Arfophilin-2; FIP4-Rab11; RAB11 family interacting protein 4 (class II); Rab11 family-interacting protein 4
Gene Aliases: ARFO2; KIAA1821; RAB11-FIP4; RAB11FIP4
UniProt ID: (Human) Q86YS3
Entrez Gene ID: (Human) 84440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.