Invitrogen
Manufacturer Code:PA539384
Catalog # PIPA539384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human RAB11FIP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arfophilin-2; FIP4-Rab11; mRab11-FIP4A; RAB11 family interacting protein 4 (class II); Rab11 family-interacting protein 4; RAB11-FIP4 RAB11 family interacting protein 4 class II
Gene Aliases: A730072L08Rik; ARFO2; KIAA1821; mKIAA1821; RAB11-FIP4; RAB11FIP4
UniProt ID: (Human) Q86YS3, (Mouse) Q8BQP8
Entrez Gene ID: (Human) 84440, (Mouse) 268451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.