Invitrogen
Manufacturer Code:PA535959
Catalog # PIPA535959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 318 and 589 of RAB11FIP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FIP4-Rab11; mRab11-FIP4A; Rab11 family-interacting protein 4; RAB11-FIP4 RAB11 family interacting protein 4 class II
Gene Aliases: A730072L08Rik; Kiaa1821; mKIAA1821; RAB11-FIP4; Rab11fip4
UniProt ID: (Mouse) Q8BQP8
Entrez Gene ID: (Mouse) 268451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.