Invitrogen
Manufacturer Code:PA539383
Catalog # PIPA539383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human RAB11FIP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arfophilin-1; CART1 Rab11-FIP3 EF hands-containing Rab-interacting protein FIP3-Rab11 MU-MB-17.148 PAC196A12.1 arfophilin-1 cytoplasmic adaptor for RAR and TR eferin rab11 family-interacting protein 3 rab11-family interacting protein 3; cytoplasmic adaptor for RAR and TR; EF hands-containing Rab-interacting protein; Eferin; FIP3-Rab11; MU-MB-17.148; PAC196A12.1; RAB11 family interacting protein 3 (class II); Rab11 family-interacting protein 3; rab11-family interacting protein 3
Gene Aliases: ARFO1; CART1; D030060O14Rik; KIAA0665; LRRGT00043; mKIAA0665; Rab11-FIP3; RAB11FIP3; RGD1308952
UniProt ID: (Human) O75154, (Mouse) Q8CHD8
Entrez Gene ID: (Human) 9727, (Mouse) 215445, (Rat) 303002
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.