Abnova
Manufacturer Code:PAB11253
Catalog # 89-114-961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to C-terminus one-third of human RAB11FIP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant RAB11FIP3.
For Research Use Only
Protein Aliases: Arfophilin-1; cytoplasmic adaptor for RAR and TR; EF hands-containing Rab-interacting protein; Eferin; FIP3-Rab11; KIAA0665; MU-MB-17.148; PAC196A12.1; RAB11 family interacting protein 3 (class II); Rab11 family-interacting protein 3; rab11-family interacting protein 3; Rab11-FIP3
Gene Aliases: ARFO1; CART1; KIAA0665; Rab11-FIP3; RAB11FIP3
UniProt ID: (Human) O75154
Entrez Gene ID: (Human) 9727
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.