Invitrogen
Manufacturer Code:PA555276
Catalog # PIPA555276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RAB11FIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NOEL1A RCP rab11-FIP1 RAB11 coupling protein Rab effector protein Rab-interacting recycling protein rab-coupling protein rab11 family-interacting protein 1; Rab effector protein; Rab-coupling protein; Rab-interacting recycling protein; RAB11 coupling protein; RAB11 family interacting protein 1 (class I); Rab11 family-interacting protein 1; Rab11-FIP1
Gene Aliases: NOEL1A; rab11-FIP1; RAB11FIP1; RCP
UniProt ID: (Human) Q6WKZ4
Entrez Gene ID: (Human) 80223
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.