Proteintech
Manufacturer Code:167781AP150UL
Catalog # 50-173-1172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAB11FIP1 Fusion Protein Ag10179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NOEL1A Rab coupling protein rab11 FIP1 RAB11FIP1 RCP; Rab coupling protein; Rab effector protein; Rab-coupling protein; Rab-interacting recycling protein; RAB11 coupling protein; RAB11 family interacting protein 1 (class I); Rab11 family-interacting protein 1; Rab11-FIP1; rab11fip1 {ECO:0000250|UniProtKB:Q6WKZ4}
Gene Aliases: 2010200K21Rik; 4833414G05Rik; NOEL1A; Rab11-FIP1; RAB11FIP1; RCP; RGD1562965
UniProt ID: (Human) Q6WKZ4, (Mouse) Q9D620, (Rat) Q3B7T9
Entrez Gene ID: (Human) 80223, (Mouse) 75767, (Rat) 498650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.