Proteintech
Manufacturer Code:197421AP150UL
Catalog # 50-173-2519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein YPT3 H YPT3 RAB11B Ras related protein Rab 11B YPT3; GTP-binding protein YPT3; RAB11B, member of RAS oncogene family; RAS oncogene family protein; Ras-related protein Rab-11B
Gene Aliases: A730055L17Rik; H-YPT3; RAB11B; YPT3
UniProt ID: (Human) Q15907, (Rat) O35509, (Mouse) P46638
Entrez Gene ID: (Human) 9230, (Rat) 79434, (Mouse) 19326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.