Invitrogen
Manufacturer Code:MA124919
Catalog # PIMA124919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7C10 |
Immunogen | Synthetic peptide corresponding to residues K Q M S D R R E N D M of Rab11A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC1490 YL8; RAB 11A, member oncogene family; Rab-11; Ras-related protein Rab-11A; YL8
Gene Aliases: RAB11; RAB11A; YL8
UniProt ID: (Human) P62491
Entrez Gene ID: (Human) 8766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.